missing translation for 'onlineSavingsMsg'
Learn More

(S)-Alpha-Hydroxy-Beta-methoxy-Beta-phenyl-benzenepropanoic Acid Methyl Ester, TRC

Product Code. 30396954
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396954 500 mg
30405575 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396954 Supplier Toronto Research Chemicals Supplier No. TRCH946295500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177036-78-1
Molecular Formula C17H18O4
Molecular Weight (g/mol) 286.32
Synonym (S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic Acid Methyl Ester, Ambrisentan Hydroxyester Impurity
IUPAC Name methyl (S)-2-hydroxy-3-methoxy-3,3-diphenylpropanoate
SMILES CoC(=O)[C@@H](O)C(OC)(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C17H18O4/c1-20-16(19)15(18)17(21-2,13-9-5-3-6-10-13)14-11-7-4-8-12-14/h3-12,15,18H,1-2H3/t15-/m1/s1
Formula Weight 286.12
Chemical Name or Material (S)-alpha-Hydroxy-beta-methoxy-beta-phenyl-benzenepropanoic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.