missing translation for 'onlineSavingsMsg'
Learn More

(S)-alpha-Allylglycine ethyl ester p-toluenesulfonate, 98%, 98% ee, Thermo Scientific™

Product Code. 11945771 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
11945771 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 11945771

Brand: Thermo Scientific Acros 441280010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 1231709-21-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 131°C to 133°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C7H13NO2·C7H8O3S
Quantity 1 g
Specific Rotation 13
Synonym s-allylglycine ethyl ester p-toluenesulfonateee, ethyl 2s-2-amino-4-pentenoate 4-methylbenzenesulfonate 1:1
InChI Key MEQLYDZSCPUCRY-RGMNGODLSA-N
SMILES CCOC(=O)C(CC=C)N.CC1=CC=C(C=C1)S(=O)(=O)O
IUPAC Name ethyl (2S)-2-aminopent-4-enoate;4-methylbenzenesulfonic acid
Molecular Weight (g/mol) 315.39
PubChem CID 119026235
Formula Weight 315.39
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (S)-α-Allylglycine ethyl ester p-toluenesulfonate, 98%
Show More Show Less
Product Identifier
  • (S)-alpha-Allylglycine ethyl ester p-toluenesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.