missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha-(1-Naphthyl)ethylamine, 99+%, Thermo Scientific Chemicals

Catalog Number p-3758543
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10420-89-0
C12H14N
172.25
MFCD00064179
RTCUCQWIICFPOD-VIFPVBQESA-O
s---1-1-naphthyl ethylamine, s-1-naphthalen-1-yl ethanamine, 1s-1-naphthalen-1-yl ethan-1-amine, s---alpha-1-naphthyl ethylamine, s-1-1-naphthyl ethylamine, 1s-1-1-naphthyl ethanamine, s---1-naphthyl ethylamine, s-alpha-methyl-1-naphthalenemethanamine, 1s-1-naphthalen-1-yl ethanamine
66325
(1S)-1-naphthalen-1-ylethanamine
C[C@H]([NH3+])C1=C2C=CC=CC2=CC=C1
This item is not returnable. View return policy
10420-89-0
C12H14N
172.25
MFCD00064179
RTCUCQWIICFPOD-VIFPVBQESA-O
s---1-1-naphthyl ethylamine, s-1-naphthalen-1-yl ethanamine, 1s-1-naphthalen-1-yl ethan-1-amine, s---alpha-1-naphthyl ethylamine, s-1-1-naphthyl ethylamine, 1s-1-1-naphthyl ethanamine, s---1-naphthyl ethylamine, s-alpha-methyl-1-naphthalenemethanamine, 1s-1-naphthalen-1-yl ethanamine
66325
(1S)-1-naphthalen-1-ylethanamine
C[C@H]([NH3+])C1=C2C=CC=CC2=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Color Orange to Yellow
Density 1.0600g/mL
Boiling Point 153.0°C (11.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CH(CH3)NH2
Refractive Index 1.6217 to 1.6237
Quantity 1 g
Beilstein 12, III, 3111
Specific Gravity 1.06
Specific Rotation Condition - 60.00 (20.00°C c=2,CH3OH)
Specific Rotation − 60.00
Formula Weight 171.24
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material (S)-(-)-α-(1-Naphthyl)ethylamine
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-a-(1-Naphthyl)ethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'