missing translation for 'onlineSavingsMsg'
Learn More

S-Adenosyl-L-methionine, MedChemExpress

Product Code. 16428396
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16428396

Brand: Medchem Express HYB0617

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress S-Adenosyl-L-methionine (S-Adenosyl methionine) is produced endogenously from methionine and ATP by action of the enzyme methionine adenosyltransferase and is an important orally active methyl group donor. S-Adenosyl-L-methionine is a dietary supplement with potent antidepressant effects, and has the potential for liver disease and osteoarthritis research.

S-Adenosyl-L-methionine (S-Adenosyl methionine) is produced endogenously from methionine and ATP by action of the enzyme methionine adenosyltransferase and is an important orally active methyl group donor. S-Adenosyl-L-methionine is a dietary supplement with potent antidepressant effects, and has the potential for liver disease and osteoarthritis research.

Specifications

Chemical Name or Material S-Adenosyl-L-methionine
CAS 29908-03-0
Color White
Purity Grade Notes Research
Synonym S-Adenosyl methionine Ademetionine AdoMet
Solubility Information DMSO : 100 mg/mL (250.98 mM; Need ultrasonic) ∣H2O : ≥ 43 mg/mL (107.92 mM)
SMILES O[C@H]1[C@](O[C@@H]([C@H]1O)C[S+](C)CC[C@H](N)C([O-])=O)([H])N2C3=NC=NC(N)=C3N=C2
Molecular Weight (g/mol) 398.44
Quantity 100 mg
Formula Weight 398.44
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.