missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-Adenosyl-L-methionine disulfate tosylate, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
Description
S-Adenosyl-L-methionine disulfate tosylate (Ademetionine disulfate tosylate) is the principal biological methyl donor synthesized in all mammalian cells but most abundantly in the liver.
Specifications
Specifications
| Chemical Name or Material | S-Adenosyl-L-methionine disulfate tosylate |
| CAS | 97540-22-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H34N6O16S4 |
| Synonym | Ademetionine disulfate tosylateS-Adenosyl methionine disulfate tosylateAdoMet disulfate tosylate |
| Solubility Information | DMSO : ≥ 50 mg/mL (65.21 mM) ∣H2O : 33.33 mg/mL (43.47 mM; Need ultrasonic) |
| SMILES | N[C@@H](CC[S+](C[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3N)O1)O)O)C)C(O)=O.O=S(O)([O-])=O.O=S(O)(O)=O.OS(=O)(C4=CC=C(C)C=C4)=O |
| Molecular Weight (g/mol) | 766.8 |
| Quantity | 100 mg |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction