missing translation for 'onlineSavingsMsg'
Learn More

(S)-7-Hydroxy Efavirenz, TRC

Product Code. 30393154
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393154 1 mg -
30395275 10 mg -
30393903 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393154 Supplier Toronto Research Chemicals Supplier No. TRCH8053451MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 252343-24-1
Molecular Formula C14H9ClF3NO3
Molecular Weight (g/mol) 331.67
Synonym (4S)-6-Chloro-4-(2-cyclopropylethynyl)-1,4-dihydro-7-hydroxy-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one
IUPAC Name (4S)-6-chloro-4-(2-cyclopropylethynyl)-7-hydroxy-4-(trifluoromethyl)-1H-3,1-benzoxazin-2-one
SMILES Oc1cc2NC(=O)O[C@@](C#CC3CC3)(c2cc1Cl)C(F)(F)F

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C14H9ClF3NO3/c15-9-5-8-10(6-11(9)20)19-12(21)22-13(8,14(16,17)18)4-3-7-1-2-7/h5-7,20H,1-2H2,(H,19,21)/t13-/m0/s1
Formula Weight 331.0223
Chemical Name or Material (S)-7-Hydroxy Efavirenz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.