missing translation for 'onlineSavingsMsg'
Learn More

(S)-6-Hydroxy Warfarin, TRC

Product Code. 30400969
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400969 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400969 Supplier Toronto Research Chemicals Supplier No. TRCH99611625MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 63740-80-7
Molecular Formula C19H16O5
Quantity 25 mg
Synonym 4,6-Dihydroxy-3-[(1S)-3-oxo-1-phenylbutyl]-2H-1-benzopyran-2-one, (S)-4,6-Dihydroxy-3-(3-oxo-1-phenylbutyl)-2H-1-benzopyran-2-one, (S)-6-Hydroxywarfarin, 6-Hydroxy-S-warfarin
InChI Formula InChI=1S/C19H16O5/c1-11(20)9-14(12-5-3-2-4-6-12)17-18(22)15-10-13(21)7-8-16(15)24-19(17)23/h2-8,10,14,21-22H,9H2,1H3/t14-/m0/s1
SMILES CC(=O)C[C@H](C1=C(O)c2cc(O)ccc2OC1=O)c3ccccc3
IUPAC Name 4,6-dihydroxy-3-[(1S)-3-oxo-1-phenylbutyl]chromen-2-one
Molecular Weight (g/mol) 324.33
Formula Weight 324.0998
Chemical Name or Material (S)-6-Hydroxy Warfarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.