missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone, 98%

Product Code. 11374975
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374975 1 g 1g
11456850 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374975 Supplier Thermo Scientific Alfa Aesar Supplier No. L18359.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17342-08-4 | C5H9NO2 | 115.132 g/mol

(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone serves as a building block in the synthesis of (R)- and (S)-diaminovaleric acids1 and of 5-azasemicorrin bidentate nitrogen ligands for enantioselective catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone serves as a building block in the synthesis of (R)- and (S)-diaminovaleric acids1 and of 5-azasemicorrin bidentate nitrogen ligands for enantioselective catalysis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17342-08-4
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00077792
InChI Key HOBJEFOCIRXQKH-BYPYZUCNSA-N
Synonym l-pyroglutaminol, s-+-5-hydroxymethyl-2-pyrrolidinone, 5s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl-2-pyrrolidinone, 2-pyrrolidinone, 5-hydroxymethyl-, 5s, s-pyroglutaminol, l-pyroglutamol, pubchem13847
PubChem CID 643511
IUPAC Name (5S)-5-(hydroxymethyl)pyrrolidin-2-one
SMILES C1CC(=O)NC1CO

Specifications

Melting Point 78°C to 82°C
Boiling Point 147°C to 149°C (0.06 mmHg)
Quantity 1 g
Beilstein 4657914
Solubility Information Soluble in water.
Optical Rotation +30° (c=2 in Ethanol)
Formula Weight 115.13
Percent Purity 98%
Chemical Name or Material (S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.