missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-5-Fluorowillardiine hydrochloride, MedChemExpress

Product Code. p-200100654
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16457158

Brand: Medchem Express HY16713A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S)-(-)-5-Fluorowillardiine hydrochloride is a potent and specific AMPAR agonist.

(S)-(-)-5-Fluorowillardiine hydrochloride is a potent and specific AMPAR agonist.

Specifications

Chemical Name or Material (S)-(-)-5-Fluorowillardiine hydrochloride
Physical Form Solid
CAS 1321546-70-6
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C7H9ClFN3O4
For Use With (Application) Neuroscience-Neuromodulation
Synonym (5S)-Fluorowillardiine hydrochloride (S)-5-Fluorowillardiine hydrochloride
Solubility Information H2O : 6.67 mg/mL (26.30 mM; Need ultrasonic)
SMILES OC([C@@H](N)CN1C=C(F)C(NC1=O)=O)=O.[H]Cl
Molecular Weight (g/mol) 253.62
Formula Weight 253.62
Percent Purity 98.06%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.