missing translation for 'onlineSavingsMsg'
Learn More

(S)-5-Chloro-3-(cyclopropylethynyl)-3-(trifluoromethyl)-1,3-dihydrobenzo[c]isoxazole, TRC

Product Code. 30367327
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367327 250 mg 250mg
30357387 50 mg 50mg
30367380 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30367327 Supplier Toronto Research Chemicals Supplier No. TRCC630765250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C14 H11 Cl F3 N O
Molecular Weight (g/mol) 301.692
IUPAC Name (4R)-6-chloro-4-(2-cyclopropylethynyl)-4-(trifluoromethyl)-1,3-dihydro-2,1-benzoxazine
SMILES FC(F)(F)[C@]1(CONc2ccc(Cl)cc12)C#CC3CC3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H11ClF3NO/c15-10-3-4-12-11(7-10)13(8-20-19-12,14(16,17)18)6-5-9-1-2-9/h3-4,7,9,19H,1-2,8H2/t13-/m0/s1
Formula Weight 301.048
Chemical Name or Material (S)-5-Chloro-3-(cyclopropylethynyl)-3-(trifluoromethyl)-1,3-dihydrobenzo[c]isoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.