missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-(5'-Adenosyl)-L-methionine-d3 Disulfate Salt (Synthetic) (Mixture of Diastereomers), TRC
Click to view available options
Quantity:
1 mg
10 mg
Specifications
Specifications
| Chemical Name or Material | S-(5'-Adenosyl)-L-methionine-d3 Disulfate Salt (Mixture of Diastereomers) |
| Molecular Formula | C15 DH3 H20 N6 O5 S . H S O4 . H2 S O4 |
| Quantity | 1 mg |
| Synonym | 5'-[[(3S)-3-Amino-3-carboxypropyl](methyl-d3)sulfonio]-5'-deoxyadenosine Disulfate, 5'-[(L-3-Amino-3-carboxypropyl)(methyl-d3)sulfonio]-5'-deoxyadenosine Disulfate, S-Adenosylmethionine-d3 Disulfate, Active Methionine-d3 Disulfate, Ademetionine-d3 Disulfate, L-S-Adenosylmethionin-d3 Disulfate, S-Adenosyl-L-methionine-d3 Disulfate |
| InChI Formula | InChI=1S/C15H22N6O5S.2H2O4S/c1-27(3-2-7(16)15(24)25)4-8-10(22)11(23)14(26-8)21-6-20-9-12(17)18-5-19-13(9)21;2*1-5(2,3)4/h5-8,10-11,14,22-23H,2-4,16H2,1H3,(H2-,17,18,19,24,25);2*(H2,1,2,3,4)/t7-,8+,10?,11-,14+,27?;;/m0../s1/i1D3;; |
| SMILES | [2H]C([2H])([2H])[S+](CC[C@H](N)C(=O)O)C[C@H]1O[C@H]([C@@H](O)C1O)n2cnc3c(N)ncnc23.OS(=O)(=O)O.OS(=O)(=O)[O-] |
| IUPAC Name | [(3S)-3-amino-3-carboxy-propyl]-[[(2S,4S,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methyl]-(trideuteriomethyl)sulfonium;hydrogen sulfate;sulfuric acid |
| Molecular Weight (g/mol) | 597.612 |
| Formula Weight | 597.0908 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction