missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-(5'-Adenosyl)-L-methionine-13C5 Sulfate Salt (Synthetic)(Mixture of Diastereomers), TRC
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
Specifications
Specifications
| Chemical Name or Material | S-(5'-Adenosyl)-L-methionine-13C5 Sulfate Salt (Mixture of Diastereomers) |
| Molecular Formula | C1013C5H23N6O5S+ •HSO4- •xH2SO4 |
| Quantity | 0.5 mg |
| Synonym | 5'-[[(3S)-3-Amino-3-carboxypropyl]methylsulfonio]-5'-deoxyadenosine-13C5 Disulfate, 5'-[(L-3-Amino-3-carboxypropyl)methylsulfonio]-5'-deoxyadenosine-13C5 Disulfate, S-Adenosylmethionine-13C5 Disulfate, Active Methionine-13C5 Disulfate, Ademetionine-13C5 Disulfate, L-S-Adenosylmethionin-13C5 Disulfate, S-Adenosyl-L-methionine-13C5 Disulfate |
| SMILES | NC1=NC=NC2=C1N=CN2[13C@H]3[13C@H](O)[13C@H](O)[13C@@H]([13CH2][S+](C)CC[C@H](N)C(O)=O)O3.[HSO4-] |
| Molecular Weight (g/mol) | 404.4197079808 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction