missing translation for 'onlineSavingsMsg'
Learn More

S 421-d4, TRC

Product Code. 30423755
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30423755 1 mg
30410028 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30423755 Supplier Toronto Research Chemicals Supplier No. TRCS0505021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C6 D4 H2 Cl8 O
Molecular Weight (g/mol) 381.76
Synonym Bis(2,3,3,3-tetrachloropropyl) Ether-d4, 2,3,3,3,2',3',3',3'-Octachlorodipropyl Ether-d4, Octachloro-di-n-propyl Ether-d4, Octachlorodipropyl Ether-d4, SV2-d4, S 421-d4
IUPAC Name 1,1,1,2-tetrachloro-3,3-dideuterio-3-(2,3,3,3-tetrachloro-1,1-dideuteriopropoxy)propane
SMILES [2H]C([2H])(OC([2H])([2H])C(Cl)C(Cl)(Cl)Cl)C(Cl)C(Cl)(Cl)Cl

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C6H6Cl8O/c7-3(5(9,10)11)1-15-2-4(8)6(12,13)14/h3-4H,1-2H2/i1D2,2D2
Formula Weight 377.818
Chemical Name or Material 1,1'-Oxybis-2,3,3,3-tetrachloropropane-d4
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.