missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-4-Penten-2-ol, 97%

Product Code. 11319939
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319939 5 g 5g
11309939 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319939 Supplier Thermo Scientific Alfa Aesar Supplier No. B25728.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55563-79-6 | C5H10O | 86.134 g/mol

(S)-(+)-4-Penten-2-ol is involved in the synthesis of goniothalamin, hexadecanolide, massoia lactone, and parasorbic acid through sequential allylboration-esterification ring-closing metathesis reactions. It is also used to prepare 2-pentanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-4-Penten-2-ol is involved in the synthesis of goniothalamin, hexadecanolide, massoia lactone, and parasorbic acid through sequential allylboration-esterification ring-closing metathesis reactions. It is also used to prepare 2-pentanone.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55563-79-6
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD03427200
InChI Key ZHZCYWWNFQUZOR-YFKPBYRVSA-N
Synonym s-+-4-penten-2-ol, 4-penten-2-ol, 2s, 2s-pent-4-en-2-ol, 2s-4penten-2-ol, s-4-pentene-2-ol, 2s-4-penten-2-ol, s-pent-4-en-2-ol, s-2-hydroxypent-4-ene
PubChem CID 6994332
IUPAC Name (2S)-pent-4-en-2-ol
SMILES CC(CC=C)O

Specifications

Density 0.837
Boiling Point 115°C to 116°C
Flash Point 30°C (86°F)
Refractive Index 1.425
Quantity 5 g
UN Number UN1987
Optical Rotation +5.2° (c=1 in Chloroform)
Formula Weight 86.13
Percent Purity 97%
Chemical Name or Material (S)-(+)-4-Penten-2-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.