missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-4-Isopropyl-2-oxazolidinone, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | (S)-(-)-4-Isopropyl-2-oxazolidinone |
| CAS | 17016-83-0 |
| Molecular Formula | C6 H11 N O2 |
| Quantity | 2.5 g |
| Synonym | 2-Oxazolidinone, 4-(1-methylethyl)-, (4S)-, 2-Oxazolidinone, 4-(1-methylethyl)-, (S)-, 2-Oxazolidinone, 4-isopropyl-, L- (8CI), (4S)-4-(1-Methylethyl)-2-oxazolidinone, (4S)-(-)-4-Isopropyl-2-oxazolidinone, (4S)-(-)-4-Isopropyl-2-oxazolidone, (4S)-2-Oxo-4-(propan-2-yl)-1,3-oxazolidine, (4S)-4-(Propan-2-yl)-1,3-oxazolidin-2-one, (4S)-4-Isopropyl-1,3-oxazolidin-2-one, (4S)-4-Isopropyl-2-oxazolidinone, (S)-(-)-4-Isopropyl-2-oxazolidinone, (S)-4-Isopropyl-2-oxazolidinone, (S)-4-Isopropyl-2-oxazolidone, (S)-4-Isopropyloxazolidin-2-one, 4(S)-(1-Methylethyl)-2-oxazolidinone, 4(S)-Isopropyloxazolidin-2-one |
| InChI Formula | InChI=1S/C6H11NO2/c1-4(2)5-3-9-6(8)7-5/h4-5H,3H2,1-2H3,(H,7,8)/t5-/m1/s1 |
| SMILES | CC(C)[C@H]1COC(=O)N1 |
| IUPAC Name | (4S)-4-propan-2-yl-1,3-oxazolidin-2-one |
| Molecular Weight (g/mol) | 129.16 |
| Formula Weight | 129.079 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction