missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Hydroxy Penbutolol Hydrochloride, TRC

Product Code. 30392509
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392509 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392509 Supplier Toronto Research Chemicals Supplier No. TRCH9489452.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-4-Hydroxy Penbutolol Hydrochloride
CAS 1346617-04-6
Molecular Formula C18H30ClNO3
Quantity 2.5 mg
Synonym 3-Cyclopentyl-4-[(2S)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenol Hydrochloride, (S)-3-Cyclopentyl-4-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenol Hydrochloride
InChI Formula InChI=1S/C18H29NO3.ClH/c1-18(2,3)19-11-15(21)12-22-17-9-8-14(20)10-16(17)13-6-4-5-7-13;/h8-10,13,15,19-21H,4-7,11-12H2,1-3H3;1H/t15-;/m0./s1
SMILES Cl.CC(C)(C)NC[C@H](O)COc1ccc(O)cc1C2CCCC2
IUPAC Name 4-[(2S)-3-(tert-butylamino)-2-hydroxypropoxy]-3-cyclopentylphenol;hydrochloride
Molecular Weight (g/mol) 343.89
Formula Weight 343.1914
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.