missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-4-Hydroxy-2-pyrrolidinone, 98%, Thermo Scientific Chemicals

Product Code. 10112904 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10112904 5 g Glass bottle 5g
10102904 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10112904 Supplier Thermo Scientific Acros Supplier No. 398970050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 68108-18-9
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.1
InChI Key IOGISYQVOGVIEU-VKHMYHEASA-N
Synonym s-4-hydroxy-2-pyrrolidinone, s-4-hydroxypyrrolidin-2-one, s-4-hydroxy-2-pyrrolidone, s-4-hydroxypyrrolidine-2-one, 4s-4-hydroxypyrrolidin-2-one, s---4-hydroxy-2-pyrrolidinone, 2-pyrrolidinone, 4-hydroxy-, 4s, s---4-hydroxy-2-pyrrolidone, s-4-hydroxy-pyrrolidine-2-one, 4s-4-hydroxy-2-pyrrolidinone
PubChem CID 155084
IUPAC Name (4S)-4-hydroxypyrrolidin-2-one
SMILES C1C(CNC1=O)O

Spezifikation

Melting Point 156°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 101.1
Percent Purity 98%
Chemical Name or Material (S)-(-)-4-Hydroxy-2-pyrrolidinone
compliance-icons
Produktidentifikator
  • (S)-(-)-4-Hydroxy-2-pyrrolidinone
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.