missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid, 96%

Product Code. 15469485
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469485 250 mg 250mg
15438487 1 g 1g
15448487 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15469485 Supplier Thermo Scientific Alfa Aesar Supplier No. H51990.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 125238-99-5 | C24H28N2O6 | 440.50 g/mol

It finds its application in combination with the Nγ-Z derivative for the synthesis of polymyxin analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in combination with the Nγ-Z derivative for the synthesis of polymyxin analogs.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125238-99-5
Molecular Formula C24H28N2O6
Molecular Weight (g/mol) 440.50
MDL Number MFCD00151941
InChI Key LIWKOFAHRLBNMG-UHFFFAOYNA-N
Synonym fmoc-dab boc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid, fmoc-n4-boc-l-2,4-diaminobutyric acid, 2s-4-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, fmoc-l-dab boc-oh, fmoc-dab boc, ambotzfaa1364, n-fmoc-n'-boc-l-2,4-diaminobutyric acid, na-fmoc-n-boc-l-2,4-diaminobutyric acid, fmoc-dab boc-oh hplc
PubChem CID 2756101
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)NCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 111°C to 113°C
Quantity 250 mg
Beilstein 6250328
Solubility Information Slightly soluble in water.
Optical Rotation −12° (c=1 in Methanol)
Formula Weight 440.5
Percent Purity 96%
Chemical Name or Material (S)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.