missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Benzyl-3-butyryl-2-oxazolidinone, Thermo Scientific™

Product Code. 15436609
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436609 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15436609 Supplier Thermo Scientific Alfa Aesar Supplier No. J64844.#0
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • A cell-permeable N-(butyryl)oxazolidinone compound that serves as a negative control for the Cell Sheet Migration Inhibitor
TRUSTED_SUSTAINABILITY

Specifications

CAS 112459-79-7
Color White
Molecular Formula C14H17NO3
MDL Number MFCD06411406
Quantity 5 mg
Synonym s-4-benzyl-3-butyryloxazolidin-2-one, 4s-3-butyryl-4-benzyl-2-oxazolidinone, s-4-benzyl-3-butyryl-2-oxazolidinone, cell sheet migration inhibitor, negative control, 4s-4-benzyl-3-butanoyl-1,3-oxazolidin-2-one, 4s-4-benzyl-3-butyryl-1,3-oxazolidin-2-one, 2-oxazolidinone, 3-1-oxobutyl-4-phenylmethyl-, s, 4s-3-butyryl-4-benzyloxazolidin-2-one, 4s-4-benzyl-3-butanoyl-oxazolidin-2-one, n-butyryl-4-s phenylmethyl-2-oxazolidinone
InChI Key HEJIYTVOUUVHRY-LBPRGKRZSA-N
SMILES CCCC(=O)N1C(COC1=O)CC2=CC=CC=C2
IUPAC Name (4S)-4-benzyl-3-butanoyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol) 247.294
PubChem CID 11010277
Formula Weight 247.29
Physical Form Powder
Chemical Name or Material (S)-4-Benzyl-3-butyryl-2-oxazolidinone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.