missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Amino-2-hydroxybutyric acid, 98%

Product Code. 11472150
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
missing translation for 'unitSize'
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472150 25 g 25g
11462150 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11472150 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' H28796.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40371-51-5 | C4H9NO3 | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40371-51-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00674110
InChI Key IVUOMFWNDGNLBJ-VKHMYHEASA-N
Synonym s-4-amino-2-hydroxybutanoic acid, s-4-amino-2-hydroxybutyric acid, s---4-amino-2-hydroxybutyric acid, 2s-4-amino-2-hydroxybutanoic acid, s---amino-2-hydroxybutyric acid, butanoic acid, 4-amino-2-hydroxy-, 2s, s---4-amino-2-hydroxybutyricacid, pubchem23215, l-2-hydroxy-4-aminobutyric acid
PubChem CID 2733929
IUPAC Name (2S)-4-amino-2-hydroxybutanoic acid
SMILES [NH3+]CC[C@H](O)C([O-])=O

Specifications

Quantity 25 g
Formula Weight 119.12
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material (S)-4-Amino-2-hydroxybutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.