missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-[4-(Oxiranylmethoxy)-1,2,5-thiadiazol-3-yl]morpholine, TRC

Product Code. 30418752
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30418752 250 mg
30420476 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30418752 Supplier Toronto Research Chemicals Supplier No. TRCO847080250MG
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69500-53-4
Molecular Formula C9 H13 N3 O3 S
Molecular Weight (g/mol) 243.28
IUPAC Name 4-[4-[[(2S)-oxiran-2-yl]methoxy]-1,2,5-thiadiazol-3-yl]morpholine
SMILES C(Oc1nsnc1N2CCOCC2)[C@@H]3CO3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H13N3O3S/c1-3-13-4-2-12(1)8-9(11-16-10-8)15-6-7-5-14-7/h7H,1-6H2/t7-/m0/s1
Formula Weight 243.0678
Chemical Name or Material (S)-4-[4-(Oxiranylmethoxy)-1,2,5-thiadiazol-3-yl]morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.