missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-(4-Hydrazinobenzyl)-2-oxazolidinone Hydrochloride, TRC

Product Code. 30390985
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390985 25 mg 25mg
30396243 5 mg -
30396549 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390985 Supplier Toronto Research Chemicals Supplier No. TRCH71649525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139264-57-6
Molecular Formula C10H13N3O2
Molecular Weight (g/mol) 243.69
Synonym (S)-4-(4-Hydrazinobenzyl)-1,3-oxazolidin-2-one Hydrochloride, (S)-4-[(4-Hydrazinophenyl)methyl]-2-oxazolidinone, (4S)-4-[(4-Hydrazinylphenyl)methyl]-2-oxazolidinone Hydrochloride
IUPAC Name (4S)-4-[(4-hydrazinylphenyl)methyl]-1,3-oxazolidin-2-one;hydrochloride
SMILES Cl.NNc1ccc(C[C@H]2COC(=O)N2)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H13N3O2.ClH/c11-13-8-3-1-7(2-4-8)5-9-6-15-10(14)12-9;/h1-4,9,13H,5-6,11H2,(H,12,14);1H/t9-;/m0./s1
Formula Weight 243.0775
Chemical Name or Material (S)-4-(4-Hydrazinobenzyl)-2-oxazolidinone Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.