missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-(4-Aminobenzyl)-2-oxazolidinone, 97%, Thermo Scientific™

Product Code. 15472438
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15472438 1 g 1g
15492438 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15472438 Supplier Thermo Scientific Alfa Aesar Supplier No. H56017.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-4-(4-Aminobenzyl)-2-(1H)-oxazolidinone is an intermediate of zolmitriptan (sc-220415) which acts as a novel inhibitor of the cytochrome P-450 enzyme aromatase.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store at -20°C. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152305-23-2
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.218
MDL Number MFCD03411476
InChI Key WNAVSKJKDPLWBD-VIFPVBQESA-N
Synonym s-4-4-aminobenzyl oxazolidin-2-one, s-4-4-aminobenzyl-2-oxazolidinone, s-4-4-aminobenzyl-2 1h-oxazolidinone, s-4-4-aminobenzyl-1,3-oxazolidin-2-one, s-4-4-amino-benzyl-oxazolidine-2-one, 4s-4-4-aminophenyl methyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-4-aminophenyl methyl-, 4s, s-4-4-aminobenzyl-1,3-oxozolidin-2-one, 4s-4-4-aminophenyl methyl-2-oxazolidinone, s-4-4'-aminobenzyl-1,3-oxazolidin-2-one
PubChem CID 7099156
IUPAC Name (4S)-4-[(4-aminophenyl)methyl]-1,3-oxazolidin-2-one
SMILES C1C(NC(=O)O1)CC2=CC=C(C=C2)N

Specifications

Melting Point 107°C to 111°C
Quantity 1 g
Solubility Information Soluble in dimethyl sulfoxide and methanol.
Formula Weight 192.22
Percent Purity 97%
Chemical Name or Material (S)-4-(4-Aminobenzyl)-2-oxazolidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.