missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-(4-Aminobenzyl)-2-(1H)-oxazolidinone, TRC

Product Code. 30010176
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010176 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30010176 Supplier Toronto Research Chemicals Supplier No. TRCA5995005G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 152305-23-2
Analyte or Component Names (S)-4-(4-Aminobenzyl)-2-(1H)-oxazolidinone
Shipping Condition Room Temperature
Molecular Formula C10 H12 N2 O2
Synonym 2-Oxazolidinone, 4-[(4-aminophenyl)methyl]-, (4S)-, 2-Oxazolidinone, 4-[(4-aminophenyl)methyl]-, (S)-, (4S)-4-[(4-Aminophenyl)methyl]-2-oxazolidinone, (4S)-4-(4-Aminobenzyl)-1,3-oxazolidin-2-one, (4S)-4-Aminophenylmethyl-oxazolidine-2-one, (S)-4-(4-Aminobenzyl)-1,3-oxazolidin-2-one, (S)-4-(4-Aminobenzyl)-2-oxazolidinone, ZTR 5, (4S)-4-(4-aminobenzyl)-1,3-oxazolidin-2-one, (4S)-4-[(4-Aminophenyl)methyl]-1,3-oxazolidin-2-one, Zolmitriptan Imp. E (EP)
InChI Formula InChI=1S/C10H12N2O2/c11-8-3-1-7(2-4-8)5-9-6-14-10(13)12-9/h1-4,9H,5-6,11H2,(H,12,13)/t9-/m0/s1
Solution Type Neat
SMILES Nc1ccc(C[C@H]2COC(=O)N2)cc1
IUPAC Name (4S)-4-[(4-aminophenyl)methyl]-1,3-oxazolidin-2-one
Industry Type Forensics
Molecular Weight (g/mol) 192.21
Quantity 5 g
Formula Weight 192.0899
Chemical Name or Material (S)-4-(4-Aminobenzyl)-2-(1H)-oxazolidinone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.