Learn More
Skytec™ S-3V Pre-Formed Cup Shape Respirator
Product Code. 30242962 Shop All Skytec Products
Change view
Click to view available options
Quantity:
5/Pk.
Unit Size:
Pack of 5
Product Code. Quantity Unit Size
30242962 5/Pk. Pack of 5
1 options
Product Code. 30242962 Supplier Skytec™ Supplier No. MMSS/ALP/S3V
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The Alpha S has been specifically designed to ensure an excellent fit for the smaller face. The unique inner face seal helps maintain a close and snug fit around the chin area which is generally regarded by experienced fit testers to be the main weakness on standard design disposable respirators.
  • Designed to solve an industry wide problem - high performance respiratory protection for the smaller face
  • Unique full inner face seal
  • Preformed nosebridge removes the need for regular adjustment
  • Four point headstrap adjustment
  • Conforms to EN 149:2001+A1:2009 FFP3 NR
Specifications
Assigned Protection Factor (APF) 20
Certifications/Compliance CE , EN 149:2000 + A1:2009 FFP3 NR
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type FFP3
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve Yes
Filtration Layer Material Melt Blown Polypropylene
Material Headstrap Textile Elastic, Woven Elastic
Model Alpha S
Quantity 5/Pk.
Sterility Non-sterile