missing translation for 'onlineSavingsMsg'
Learn More

S 3304, MedChemExpress

Product Code. 16496797
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496797 5 mg 5mg
16491069 1 mg 1mg
16472946 25 mg 25mg
16464098 10 mg 10mg
16440457 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496797 Supplier Medchem Express Supplier No. HY106992
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress S 3304 is a novel matrix metalloproteinases (MMP) inhibitor specific for MMP-2 and MMP-9.

S 3304 is a novel matrix metalloproteinases (MMP) inhibitor specific for MMP-2 and MMP-9.

Specifications

Chemical Name or Material S 3304
CAS 203640-27-1
Color Brownish Yellow
Purity Grade Notes Research
Molecular Formula C24H20N2O4S2
Quantity 5 mg
Solubility Information DMSO : ≥ 83.3 mg/mL (179.31 mM)
SMILES O=S(N[C@@H](C(O)=O)CC1=CNC2=C1C=CC=C2)(C3=CC=C(C#CC4=CC=C(C)C=C4)S3)=O
Molecular Weight (g/mol) 464.56
Formula Weight 464.56
Percent Purity 98.56%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.