Learn More
(S)-(+)-3-Methyl-2-butanol, 99%
Description
It is an pharmaceutical and organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is an pharmaceutical and organic intermediate.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation. Protect from electrostatic charges.
Specifications
Specifications
| Density | 0.81 |
| Boiling Point | 112°C |
| Flash Point | 26°C (78°F) |
| Refractive Index | 1.409 |
| Quantity | 100 mg |
| UN Number | UN1105 |
| Beilstein | 1718801 |
| Solubility Information | Soluble in alcohol. Insoluble in water. |
| Optical Rotation | +5° (Neat) |
| Formula Weight | 88.15 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.