missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Hydroxytetrahydrofuran, 99%

Product Code. 10183491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10183491 1 g Glass bottle 1g
10153591 5 g Glass bottle 5g
10580594 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10183491 Supplier Thermo Scientific Acros Supplier No. 304430010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 86087-23-2 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86087-23-2
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00064327
InChI Key XDPCNPCKDGQBAN-BYPYZUCNSA-N
Synonym s-+-3-hydroxytetrahydrofuran, s-tetrahydrofuran-3-ol, s-3-hydroxytetrahydrofuran, 3s-oxolan-3-ol, s-+-tetrahydro-3-furanol, s-tetrahydro-3-furanol, s-+-tetrahyro-3-furanol, 3s-tetrahydrofuran-3-ol, s-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3s
PubChem CID 2733227
IUPAC Name (3S)-oxolan-3-ol
SMILES C1COCC1O

Specifications

CAS Min % 98.5
Color Colorless to Yellow
CAS Max % 100.0
Density 1.2980g/mL
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4490 to 1.4510
Assay Percent Range 98.5% min. (GC)
Specific Gravity 1.298
Boiling Point 80.0°C (15.0 mmHg)
Solubility Information Solubility in water: soluble in water.
Formula Weight 88.11
Percent Purity 99%
Chemical Name or Material (S)-(+)-3-Hydroxytetrahydrofuran
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.