missing translation for 'onlineSavingsMsg'
Learn More

S-(3-Hydroxypropyl)-L-cysteine, TRC

Product Code. 30390261
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390261 100 mg -
30401609 250 mg 250mg
30393554 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390261 Supplier Toronto Research Chemicals Supplier No. TRCH952575100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13189-98-5
Molecular Formula C6 H13 N O3 S
Molecular Weight (g/mol) 179.24
Synonym Fudosteine
IUPAC Name (2R)-2-amino-3-(3-hydroxypropylsulfanyl)propanoic acid
SMILES N[C@@H](CSCCCO)C(=O)O

Specifications

InChI Formula InChI=1S/C6H13NO3S/c7-5(6(9)10)4-11-3-1-2-8/h5,8H,1-4,7H2,(H,9,10)/t5-/m0/s1
Quantity 100 mg
Formula Weight 179.0616
Shipping Condition Room Temperature
Chemical Name or Material S-(3-Hydroxypropyl)cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.