missing translation for 'onlineSavingsMsg'
Learn More

(S)-3' '-Hydroxy Pravastatin Sodium Salt, TRC

Product Code. 30396252
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396252 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396252 Supplier Toronto Research Chemicals Supplier No. TRCH9522905MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3R,5R)-3,5-Dihydroxy-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-8-[((2S,3S)3-hydroxy-2-methylbutanoyl)oxy]-2-methyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]heptanoic Acid Sodium Salt
CAS 722504-45-2
Molecular Formula C23 H35 O8 . Na
Quantity 5 mg
Synonym 1-Naphthaleneheptanoic acid, 1,2,6,7,8,8a-hexahydro-β,Δ,6-trihydroxy-8-[(2S,3S)-3-hydroxy-2-methyl-1-oxobutoxy]-2-methyl-, sodium salt (1:1), (βR,ΔR,1S,2S,6S,8S,8aR)-, 1-Naphthaleneheptanoic acid, 1,2,6,7,8,8a-hexahydro-β,Δ,6-trihydroxy-8-[(2S,3S)-3-hydroxy-2-methyl-1-oxobutoxy]-2-methyl-, monosodium salt, (βR,ΔR,1S,2S,6S,8S,8aR)- (9CI), (3R,5R)-3,5-dihydroxy-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-8-[[(2S,3S)3-hydroxy-2-methylbutanoyl]oxy]-2-methyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]heptanoic acid (3''-(S)-hydroxypravastatin), Pravastatin Sodium Imp. E (EP) as Sodium Salt
InChI Formula InChI=1S/C23H36O8.Na/c1-12-4-5-15-8-17(26)10-20(31-23(30)13(2)14(3)24)22(15)19(12)7-6-16(25)9-18(27)11-21(28)29;/h4-5,8,12-14,16-20,22,24-27H,6-7,9-11H2,1-3H3,(H,28,29);/q;+1/p-1/t12-,13-,14-,16+,17+,18+,19-,20-,22-;/m0./s1
SMILES [Na+].C[C@H](O)[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)[O-])[C@@H]12
IUPAC Name sodium;(3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-8-[(2S,3S)-3-hydroxy-2-methylbutanoyl]oxy-2-methyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
Molecular Weight (g/mol) 462.509
Formula Weight 462.223
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.