missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Hydroxy Myristic Acid Methyl Ester, TRC

Product Code. 30393409
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393409 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393409 Supplier Toronto Research Chemicals Supplier No. TRCH94851250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material S-(3)-Hydroxymyristic Acid, Methyl Ester
CAS 76835-67-1
Molecular Formula C15 H30 O3
Quantity 50 mg
InChI Formula InChI=1S/C15H30O3/c1-3-4-5-6-7-8-9-10-11-12-14(16)13-15(17)18-2/h14,16H,3-13H2,1-2H3/t14-/m0/s1
SMILES CCCCCCCCCCC[C@H](O)CC(=O)OC
IUPAC Name methyl (3S)-3-hydroxytetradecanoate
Molecular Weight (g/mol) 258.4
Formula Weight 258.2195
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.