missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(4-n-Heptylcyclohexyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 15441019
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15441019

Brand: Thermo Scientific Alfa Aesar H64005.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101532-38-1
Molecular Formula C19H29Br
Molecular Weight (g/mol) 337.345
MDL Number MFCD09838992
InChI Key OKIVMWBXCXBOGL-UHFFFAOYSA-N
Synonym 1-bromo-4-trans-4-heptylcyclohexyl benzene, 1-bromo-4-4-heptylcyclohexyl benzene, 1-bromo-4-1s,4r-4-heptylcyclohexyl benzene
PubChem CID 15135337
IUPAC Name 1-bromo-4-(4-heptylcyclohexyl)benzene
SMILES CCCCCCCC1CCC(CC1)C2=CC=C(C=C2)Br

Specifications

Quantity 5 g
Beilstein 4748130
Formula Weight 302.46
Percent Purity 98%
Physical Form Solid
Chemical Name or Material trans-4-(4-n-Heptylcyclohexyl)benzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.