missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-3-Chloro-1-phenyl-1-propanol (>90%), TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | (S)-(-)-3-Chloro-1-phenyl-1-propanol |
| CAS | 100306-34-1 |
| Molecular Formula | C9 H11 Cl O |
| Quantity | 500 mg |
| Synonym | (alphaS)-alpha-(2-Chloroethyl)benzenemethanol, alpha-(2-Chloroethyl)benzenemethanol |
| InChI Formula | InChI=1S/C9H11ClO/c10-7-6-9(11)8-4-2-1-3-5-8/h1-5,9,11H,6-7H2/t9-/m0/s1 |
| SMILES | [C@@H](CCCl)(O)C1=CC=CC=C1 |
| IUPAC Name | (1S)-3-chloro-1-phenylpropan-1-ol |
| Molecular Weight (g/mol) | 170.64 |
| Formula Weight | 170.0498 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction