missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-Butyn-2-ol, 99%

Product Code. 11309196
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309196 1 g 1g
11329196 250 mg 250mg
11319196 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11309196 Supplier Thermo Scientific Alfa Aesar Supplier No. L20345.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2914-69-4 | C4H6O | 70.091 g/mol

(S)-(-)-3-Butyn-2-ol is used to prepare (S)-(-)-2-tert-butyldimethylsiloxybut-3-yne. It finds application in ruthenium-catalyzed polymerization to get helical polyacetylenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-3-Butyn-2-ol is used to prepare (S)-(-)-2-tert-butyldimethylsiloxybut-3-yne. It finds application in ruthenium-catalyzed polymerization to get helical polyacetylenes.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2914-69-4
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00190166
InChI Key GKPOMITUDGXOSB-BYPYZUCNSA-N
Synonym s---3-butyn-2-ol, s-3-butyn-2-ol, 2s-but-3-yn-2-ol, s-but-3-yn-2-ol, 3-butyn-2-ol, 2s, s---1-butyn-3-ol, 2s-3-butyn-2-ol, 3r-+-3-hydroxybut-1-yne, s---3-butyn-2-ol, purum gc
PubChem CID 6995470
IUPAC Name (2S)-but-3-yn-2-ol
SMILES CC(C#C)O

Specifications

Density 0.891
Boiling Point 108°C to 109°C
Flash Point 28°C (82°F)
Refractive Index 1.426
Quantity 1 g
UN Number UN2929
Beilstein 4652597
Solubility Information Miscible with water.
Optical Rotation −47° (Neat)
Formula Weight 70.09
Percent Purity 99%
Chemical Name or Material (S)-(-)-3-Butyn-2-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.