missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Boc-thiazolidine-2-carboxylic acid, 97%

Product Code. p-7035087
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15429547

Brand: Thermo Scientific Alfa Aesar H52793.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 891192-95-3 | C9H15NO4S | 233.282 g/mol

(S)-3-Boc-thiazolidine-2-carboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Boc-thiazolidine-2-carboxylic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 891192-95-3
Molecular Formula C9H15NO4S
Molecular Weight (g/mol) 233.282
MDL Number MFCD02682342
InChI Key HYAXPNDMEODKHI-LURJTMIESA-N
Synonym s-3-boc-2-thiazolidinecarboxylic acid, s-3-tert-butoxycarbonyl thiazolidine-2-carboxylic acid, 2s-3-tert-butoxycarbonyl-1,3-thiazolidine-2-carboxylic acid, 2,3-thiazolidinedicarboxylic acid, 3-1,1-dimethylethyl ester, 2s, s-3-boc-thiazolidine-2-carboxylic acid, n-boc s thiazolidine-2-carboxylic acid, n-boc-s-thiazolidine-2-carboxylic acid, s-thiazolidine-2,3-dicarboxylic acid 3-tert-butyl ester, 2s-3-tert-butoxy carbonyl-1,3-thiazolidine-2-carboxylic acid, 2s-2,3-thiazolidinedicarboxylic acid, 3-1,1-dimethylethyl ester
PubChem CID 14775612
IUPAC Name (2S)-3-[(2-methylpropan-2-yl)oxycarbonyl]-1,3-thiazolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCSC1C(=O)O

Specifications

Melting Point 90°C to 93°C
Quantity 250 mg
Optical Rotation −47° (c=5 in Methanol)
Formula Weight 233.28
Percent Purity 97%
Chemical Name or Material (S)-3-Boc-thiazolidine-2-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.