missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)piperidine, TRC

Product Code. 30348414
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348414 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348414 Supplier Toronto Research Chemicals Supplier No. TRCB6211302.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-3-(Boc-amino)piperidine
CAS 216854-23-8
Molecular Formula C10 H20 N2 O2
Quantity 2.5 g
Synonym Carbamic acid, N-(3S)-3-piperidinyl-, 1,1-dimethylethyl ester, Carbamic acid, (3S)-3-piperidinyl-, 1,1-dimethylethyl ester (9CI), (3S)-(-)-3-(tert-Butoxycarbonylamino)piperidine, (3S)-3-[(tert-Butoxycarbonyl)amino]piperidine, (S)-(-)-3-(tert-Butoxycarbonylamino)piperidine, (S)-3-tert-Butoxycarbonylaminopiperidine, (S)-N-[Piperidin-3-yl]carbamic acid tert-butyl ester, (S)-Piperidine-3-ylcarbamic acid tert-butyl ester, (S)-[Piperidin-3-yl]carbamic acid tert-butyl ester, (S)-tert-Butyl (piperidin-3-yl)carbamate, (S)-tert-Butyl N-[piperidin-3-yl]carbamate, (S)-tert-Butyl [piperidin-3-yl]carbamate, tert-Butyl (3S)-piperidin-3-ylcarbamate, tert-Butyl (S)-N-[3-piperidyl]carbamate, tert-Butyl (S)-piperidin-3-ylcarbamate, (3S)-3-(tert-Butoxycarbonylamino)piperidine
InChI Formula InChI=1S/C10H20N2O2/c1-10(2,3)14-9(13)12-8-5-4-6-11-7-8/h8,11H,4-7H2,1-3H3,(H,12,13)/t8-/m0/s1
SMILES CC(C)(C)OC(=O)N[C@H]1CCCNC1
IUPAC Name tert-butyl N-[(3S)-piperidin-3-yl]carbamate
Molecular Weight (g/mol) 200.28
Formula Weight 200.1525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.