missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)piperidine, 97%

Product Code. 15413577
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413577 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15413577 Supplier Thermo Scientific Alfa Aesar Supplier No. H26957.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 216854-23-8 | C10H20N2O2 | 200.28 g/mol

(S)-3-(Boc-amino)piperidine, is used as an intermediate for pigments. It is also used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also an useful intermediate for the synthesis of a variety of chiral aminopiperidinyl quinolones as potent antibacterial agents against resistant pathogens; and alkynylpyrimidine amide derivatives as orally available inhibitors of Tie-2 kinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-(Boc-amino)piperidine, is used as an intermediate for pigments. It is also used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also an useful intermedia
TRUSTED_SUSTAINABILITY

Specifications

CAS 216854-23-8
Molecular Formula C10H20N2O2
MDL Number MFCD03093383
Synonym s-3-boc-aminopiperidine, s-3-n-boc-aminopiperidine, s-tert-butyl piperidin-3-ylcarbamate, s-3-boc-amino piperidine, tert-butyl n-3 s-piperidin-3-yl carbamate
Solubility Information Freely soluble in ethanol.
InChI Key WUOQXNWMYLFAHT-UHFFFAOYNA-N
SMILES CC(C)(C)OC(=O)NC1CCCNC1
IUPAC Name tert-butyl N-(piperidin-3-yl)carbamate
Molecular Weight (g/mol) 200.28
Quantity 5 g
PubChem CID 1514171
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material (S)-3-(Boc-amino)piperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.