missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)-4-phenylbutyric acid, 95%

Product Code. 15498717
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15498717

Brand: Thermo Scientific Alfa Aesar H52174.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51871-62-6 | C15H21NO4 | 279.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51871-62-6
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD01076271
InChI Key ACKWQHCPHJQANL-LBPRGKRZSA-N
Synonym s-3-boc-amino-4-phenylbutyric acid, boc-l-beta-homophenylalanine, boc-beta-hophe-oh, 3s-3-tert-butoxycarbonyl amino-4-phenylbutanoic acid, s-3-tert-butoxycarbonyl amino-4-phenylbutanoic acid, 3r-3-tert-butoxy carbonyl amino-4-phenylbutanoic acid, boc-hophe-oh, pubchem12107, tmba023, 3s-n-boc-3-benzyl-beta-alanine
PubChem CID 2761538
IUPAC Name (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC(O)=O)CC1=CC=CC=C1

Specifications

Melting Point 101°C to 107°C
Beilstein 3060457
Quantity 1 g
Formula Weight 279.34
Optical Rotation −19.0° (c=1 in Dichloromethane)
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)-4-phenylbutyric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.