missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)-3-phenylpropionic acid, 95%

Product Code. 15448597
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448597 5 g 5g
15458597 250 mg 250mg
15438597 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448597 Supplier Thermo Scientific Alfa Aesar Supplier No. H52072.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103365-47-5 | C14H19NO4 | 265.309 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103365-47-5
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD01860892
InChI Key JTNQFJPZRTURSI-NSHDSACASA-N
Synonym boc-s-3-amino-3-phenylpropionic acid, s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, s-3-n-boc-amino-beta-phenylalanine, s-n-boc-3-amino-3-phenylpropanoic acid, boc-d-beta-phe-oh, boc-d-beta-phenylalanine, s-boc-beta3-phenylalanine, boc-s-phenylalanine, 3s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, s-3-boc-amino-3-phenylpropionic acid
PubChem CID 2734490
IUPAC Name (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1

Specifications

Quantity 5 g
Beilstein 4695945
Formula Weight 265.31
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)-3-phenylpropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.