missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Aminopiperidine dihydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11312809
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312809 1 g 1g
15457715 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312809 Supplier Thermo Scientific Alfa Aesar Supplier No. H27700.03
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 334618-07-4 | Molecular Formula: C5H14Cl2N2 | Molecular Weight: 173.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334618-07-4
Molecular Formula C5H14Cl2N2
Molecular Weight (g/mol) 173.08
MDL Number MFCD03427036
InChI Key GGPNYXIOFZLNKW-UHFFFAOYNA-N
Synonym s-3-aminopiperidine dihydrochloride, s-piperidin-3-amine dihydrochloride, s-+-3-aminopiperidine dihydrochloride, s-3-aminopiperidine 2hcl, 3s-piperidin-3-amine dihydrochloride, s-+-3-aminopiperidine 2hcl, 3-piperidinamine, dihydrochloride, 3s, 3s-3-piperidinamine dihydrochloride, 3s-3-aminopiperidine dihydrochloride, s-piperidin-3-ylamine dihydrochloride
PubChem CID 16218278
IUPAC Name (3S)-piperidin-3-amine;dihydrochloride
SMILES Cl.Cl.NC1CCCNC1

Specifications

Melting Point ∽210°C (decomposition)
Quantity 1 g
Sensitivity Hygroscopic
Optical Rotation +1.6° (c=1 in Water)
Formula Weight 173.08
Percent Purity 98%
Chemical Name or Material (S)-(+)-3-Aminopiperidine dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.