missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Aminopiperidin-2-one, TRC

Product Code. 30009913
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009913 250 mg 250mg
30133391 500 mg 500mg
30003948 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009913 Supplier Toronto Research Chemicals Supplier No. TRCA633018250MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34294-79-6
Molecular Formula C5 H10 N2 O
Synonym (3S)-3-Amino-2-piperidinone∣(3S)-3-Aminopiperidin-2-one∣L-Orinithine Lactam;

Specifications

Quantity 250 mg
Shipping Condition Room Temperature
Formula Weight 114.0793
Molecular Weight (g/mol) 114.15
Chemical Name or Material (S)-3-Aminopiperidin-2-one
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.