missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-Amino-1,2-propanediol, 98%

Product Code. 11302739
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302739 1 g 1g
11312739 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302739 Supplier Thermo Scientific Alfa Aesar Supplier No. H26743.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 61278-21-5 | C3H9NO2 | 91.11 g/mol

(S)-(-)-3-Amino-1,2-propanediol is used as a chiral selector ligand, used subsequently for a nucleophilic substitution with the dimethoxytrityl (DMT)-protected

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-3-Amino-1,2-propanediol is used as a chiral selector ligand, used subsequently for a nucleophilic substitution with the dimethoxytrityl (DMT)-protected

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61278-21-5
Molecular Formula C3H9NO2
Molecular Weight (g/mol) 91.11
MDL Number MFCD00798260
InChI Key KQIGMPWTAHJUMN-VKHMYHEASA-N
Synonym s-3-amino-1,2-propanediol, s-3-aminopropane-1,2-diol, s---3-amino-1,2-propanediol, 2s-3-aminopropane-1,2-diol, s---3-amino-1,2-propandiol, 1,2-propanediol, 3-amino-, 2s, ksc496i7p, s-2,3-dihydroxypropylamine, 2s-2,3-dihydroxypropylamine, 2s-3-amino-1,2-propanediol
PubChem CID 440902
ChEBI CHEBI:1456
IUPAC Name (2S)-3-aminopropane-1,2-diol
SMILES C(C(CO)O)N

Specifications

Melting Point 54°C to 56°C
Density 1.175
Boiling Point 117°C to 119°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.483
Quantity 1 g
UN Number UN3259
Beilstein 1719122
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Optical Rotation −28° (c=4, 5M HCl)
Formula Weight 91.11
Percent Purity 98%
Chemical Name or Material (S)-(-)-3-Amino-1,2-propanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.