missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (S)-3,4-DCPG
GREENER_CHOICE

Product Code. 18085392
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18085392 50 mg 50mg
18084622 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085392 Supplier Tocris Bioscience™ Supplier No. 1302/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective mGlu8a agonist

Potent, selective mGlu8a agonist (EC50 = 31 nM). Displays > 100-fold selectivity over mGlu1-7 and displays little or no activity at NMDA and kainate receptors. Increases c-Fos expression in stress-related brain areas following systemic administration in mice in vivo. Also potent anticonvulsant in mice in vivo.

Caged (S)-3,4-DCPG, Racemate and R-enantiomer also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201730-11-2
Molecular Formula C10H9NO6

Specifications

Quantity 50 mg
Formula Weight 239.18
Purity >98%
Molecular Weight (g/mol) 239.183
Chemical Name or Material (S)-3, 4-DCPG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.