missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (S)-3,4-DCPG

Description
Potent, selective mGlu8a agonist (EC50 = 31 nM). Displays > 100-fold selectivity over mGlu1-7 and displays little or no activity at NMDA and kainate receptors. Increases c-Fos expression in stress-related brain areas following systemic administration in mice in vivo. Also potent anticonvulsant in mice in vivo.
Caged (S)-3,4-DCPG, Racemate and R-enantiomer also available.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 239.18 |
| Purity | >98% |
| Molecular Weight (g/mol) | 239.183 |
| Chemical Name or Material | (S)-3, 4-DCPG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?