missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 99%

Product Code. 11398046
Click to view available options
Quantity:
500 mg
2 g
Unit Size:
2g
500mg
This item is not returnable. View return policy

Product Code. 11398046

Brand: Thermo Scientific Alfa Aesar L13334.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33878-99-8 | C11H14O2 | 178.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33878-99-8
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00213797
InChI Key DNHDRUMZDHWHKG-NSHDSACASA-N
Synonym s-+-3,4,8,8a-tetrahydro-8a-methyl-1,6 2h,7h-naphthalenedione, 8as-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-3,4,7,8-tetrahydro-2h-naphthalene-1,6-dione, s-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-1,2,3,4,6,7,8,8a-octahydronaphthalene-1,6-dione, 8as-+-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 1,6 2h,7h-naphthalenedione, 3,4,8,8a-tetrahydro-8a-methyl-, 8as, wieland-mieschler ketone, 9-s-methyl-.delta.-5 10-octalin-1,6-dione, 8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione #
PubChem CID 736069
IUPAC Name (8aS)-8a-methyl-3,4,7,8-tetrahydro-2H-naphthalene-1,6-dione
SMILES CC12CCC(=O)C=C1CCCC2=O

Specifications

Melting Point 48°C to 52°C
Quantity 2 g
Beilstein 1569979
Optical Rotation +100° (c=1 in Benzene)
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material (S)-(+)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.