missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-[(2RS)-2-Amino-2-carboxyethyl]-L-homocysteine-3,3,4,4-d4, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | S-(2-Amino-2-carboxyethyl)-L-homocysteine-3,3,4,4-D4 |
| Molecular Formula | C7 H10 D4 N2 O4 S |
| Quantity | 0.01 g |
| Synonym | S-[(2 RS)-2-Amino-2-carboxyethyl]-L-homocysteine-3,3,4,4-d4, (2 S)-2-Amino-4-[(2-amino-2-carboxyethyl)thio]butanoic-3,3,4,4-d4 acid, S-(2-Amino-2-carboxyethyl)-L-homocysteine-3,3,4,4-D4 |
| InChI Formula | InChI=1 S/C7H14N2O4S/c8-4(6(10)11)1-2-14-3-5(9)7(12)13/h4-5 H,1-3,8-9H2,(H,10,11)(H,12,13)/t4-,5?/m0/s1/i1D2,2D2 |
| SMILES | [2 H]C([2 H])(SCC(N)C(=O)O)C([2 H])([2 H])[C@H](N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-4-(2-amino-2-carboxy-ethyl)sulfanyl-3,3,4,4-tetradeuterio-butanoic acid |
| Molecular Weight (g/mol) | 226.2867 |
| Formula Weight | 226.0925 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction