missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ S 2101
GREENER_CHOICE

Product Code. 18797668
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18797668

Brand: Tocris Bioscience™ 5714/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LSD1 inhibitor

S 2101 is a LSD1 inhibitor (IC50 = 990 nM; Ki = 610 nM). Exhibits selectivity for LSD1 over MAO-B and MAO-A (Ki values are 17 and 110 μM, respectively).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1R,2S)-rel-2-[3,5-Difluoro-2-(phenylmethoxy)phenyl]cycloprpanamine hydrochloride
CAS 1239262-36-2
Quantity 10 mg
Target Histone Demethylase Inhibitors
Molecular Formula C16H15F2NO.HCl
Formula Weight 311.75
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.