missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-(Trifluoromethyl)pyrrolidine, 95%

Product Code. 15493317
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493317 250 mg 250mg
11495590 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493317 Supplier Thermo Scientific Alfa Aesar Supplier No. H25854.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119580-41-5 | C5H8F3N | 139.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119580-41-5
Molecular Formula C5H8F3N
Molecular Weight (g/mol) 139.121
MDL Number MFCD03093766
InChI Key VINAMCOZNJHNIH-BYPYZUCNSA-N
PubChem CID 2782840
IUPAC Name (2S)-2-(trifluoromethyl)pyrrolidine
SMILES C1CC(NC1)C(F)(F)F

Specifications

Melting Point 27°C to 31°C
Boiling Point 110°C to 112°C
Flash Point 26°C (79°F)
Quantity 250 mg
UN Number UN2925
Sensitivity Air Sensitive
Synonym s-2-trifluoromethyl pyrrolidine, 2s-2-trifluoromethyl pyrrolidine, s-2-trifluoromethylpyrrolidine, 2 s-2-trifluoromethylpyrrolidine, s-+-2-trifluoromethyl pyrrolidine, s-2-trifluoromethyl-pyrrolidine, pyrrolidine, 2-trifluoromethyl-, s, 2s-2-trifluoromethylpyrrolidine, 2alpha-trifluoromethyl pyrrolidine, 2 s-2-trifluoromethyl pyrrolidine
Optical Rotation +10° (c=1 in Methanol)
Formula Weight 139.12
Percent Purity 95%
Chemical Name or Material (S)-(+)-2-(Trifluoromethyl)pyrrolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.