missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Trifluoromethyl)piperidine, 97%, Thermo Scientific Chemicals

£113.00 - £518.00

Chemical Identifiers

CAS 154727-51-2
Molecular Formula C6H10F3N
Molecular Weight (g/mol) 153.15
InChI Key NAXDEFXCCITWEU-YFKPBYRVSA-N
Synonym s-2-trifluoromethyl piperidine, 2s-2-trifluoromethyl piperidine, s-2-trifluoromethylpiperidine, 2alpha-trifluoromethyl piperidine
PubChem CID 42580051
IUPAC Name (2S)-2-(trifluoromethyl)piperidine
SMILES C1CCNC(C1)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11936481
View Documents Promotion Details
Thermo Scientific Acros
446650010
1 g
£113.00
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11946481
View Documents Promotion Details
Thermo Scientific Acros
446650050
5 g
£518.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

154727-51-2
153.15
s-2-trifluoromethyl piperidine, 2s-2-trifluoromethyl piperidine, s-2-trifluoromethylpiperidine, 2alpha-trifluoromethyl piperidine
(2S)-2-(trifluoromethyl)piperidine
C6H10F3N
NAXDEFXCCITWEU-YFKPBYRVSA-N
42580051
C1CCNC(C1)C(F)(F)F

Specifications

154727-51-2
96% min. (GC)
C6H10F3N
NAXDEFXCCITWEU-YFKPBYRVSA-N
(2S)-2-(trifluoromethyl)piperidine
42580051
97%
80.0°C (200.0 Torr)
Glass Bottle
s-2-trifluoromethyl piperidine, 2s-2-trifluoromethyl piperidine, s-2-trifluoromethylpiperidine, 2alpha-trifluoromethyl piperidine
C1CCNC(C1)C(F)(F)F
153.15
153.15
(S)-2-(Trifluoromethyl)piperidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

Product Identifier
  • (S)-2-(Trifluoromethyl)piperidine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only