missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-[(tert-Butoxycarbonyl)amino]-5-(dimethylamino)pentanoic acid, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 260.33 |
Percent Purity | 97% |
Chemical Name or Material | (S)-2-[(tert-Butoxycarbonyl)amino]-5-(dimethylamino)pentanoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'